Assay Details
| Gene Symbol: Kcna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173095.3 | NP_775118.1 | 1 | 1420 | 77 | |
| GenBank mRNA | M26161.1 | - | 1 | 1420 | 77 | |
| X12589.1 | - | 1633 | 77 | |||
More Information
Set Membership: |
|

Kcna1
Genomic Map