Assay Details
| Gene Symbol: Aurkaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004237.1 | NP_001004237.1 | 1 - 2 | 49 | 100 | |
| GenBank mRNA | BC079171.1 | - | 1 - 2 | 49 | 7129954 | 100 |
| FQ228270.1 | - | 1 - 2 | 57 | 100 | ||
| FQ232591.1 | - | 1 - 2 | 57 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079171.1, NM_001004237.1 transcripts.
Set Membership: |
|

Aurkaip1
Genomic Map