Assay Details
| Gene Symbol: Fam101b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007611.1 | NP_001007612.1 | 1 - 2 | 325 | 66 | |
| GenBank mRNA | BC079330.1 | - | 1 - 2 | 325 | 7096482 | 66 |
More Information
Set Membership: |
|

Fam101b
Genomic Map