Assay Details
| Gene Symbol: Lrrc8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024782.1 | NP_001019953.1 | 3 - 4 | 2326 | 72 | |
| XM_006233814.2 | XP_006233876.1 | 1 - 2 | 2172 | 72 | ||
| XM_008761660.1 | XP_008759882.1 | 1 - 2 | 2172 | 72 | ||
| GenBank mRNA | BC097371.1 | - | 3 - 4 | 2326 | 7444090 | 72 |
More Information
Set Membership: |
|

Lrrc8a
Genomic Map