Assay Details
Gene Symbol: Syngr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025644.1 | NP_001020815.1 | 3 - 4 | 552 | 77 | |
XM_006229034.2 | XP_006229096.1 | 3 - 4 | 768 | 77 | ||
GenBank mRNA | BC098938.1 | - | 3 - 4 | 552 | 7455020 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
