Assay Details
| Gene Symbol: Prl5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044702.1 | NP_001038167.1 | 1 - 2 | 93 | 109 | |
| GenBank mRNA | DQ329280.1 | - | 1 - 2 | 93 | 109 | |
More Information
Set Membership: |
|

Prl5a2
Genomic Map