Assay Details
| Gene Symbol: Mrps12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106239.1 | NP_001099709.1 | 2 - 3 | 108 | 107 | |
| XM_006228651.1 | XP_006228713.1 | 2 - 3 | 184 | 107 | ||
| XM_006228652.2 | XP_006228714.1 | 2 - 3 | 157 | 107 | ||
More Information
Set Membership: |
|

Mrps12
Genomic Map