Assay Details
| Gene Symbol: Hbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008890.1 | NP_001008890.1 | 2 - 3 | 316 | 70 | |
| GenBank mRNA | BC157809.1 | - | 2 - 3 | 322 | 9033183 | 70 |
| FQ221239.1 | - | 2 - 3 | 372 | 70 | ||
| FQ222123.1 | - | 2 - 3 | 371 | 70 | ||
More Information
Set Membership: |
|

Hbe1
Genomic Map