Assay Details
| Gene Symbol: Gpr65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106751.2 | NP_001100221.1 | 1 | 921 | 102 | |
| XM_008764761.1 | XP_008762983.1 | 2 | 1429 | 102 | ||
| GenBank mRNA | EF405799.1 | - | 1 | 921 | 102 | |
More Information
Set Membership: |
|

Gpr65
Genomic Map