Assay Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107198.1 | NP_001100668.1 | 10 - 11 | 1410 | 60 | |
| XM_008769787.1 | XP_008768009.1 | 8 - 9 | 1254 | 60 | ||
More Information
Set Membership: |
|

Exo1
Genomic Map