Assay Details
Gene Symbol: Zfp524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108905.1 | NP_001102375.1 | 1 | 922 | 81 | |
XM_008758958.1 | XP_008757180.1 | 2 | 1020 | 81 | ||
XM_008758959.1 | XP_008757181.1 | 2 | 948 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
