Assay Details
| Gene Symbol: Mum1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109321.1 | NP_001102791.1 | 1 | 2013 | 74 | |
| XM_006257301.2 | XP_006257363.1 | 5 | 2675 | 74 | ||
| XM_006257302.2 | XP_006257364.1 | 6 | 2625 | 74 | ||
| XM_006257303.2 | XP_006257365.1 | 5 | 2955 | 74 | ||
| XM_006257304.2 | XP_006257366.1 | 4 | 2898 | 74 | ||
| XM_006257305.2 | XP_006257367.1 | 5 | 2571 | 74 | ||
| GenBank mRNA | BC168654.1 | - | 3 | 2465 | 7105971 | 74 |
More Information
Set Membership: |
|

Mum1l1
Genomic Map