Assay Details
| Gene Symbol: Krt81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008814.1 | NP_001008814.1 | 8 - 9 | 1284 | 82 | |
| XM_008765774.1 | XP_008763996.1 | 8 - 9 | 1330 | 82 | ||
| GenBank mRNA | AB355638.1 | - | 8 - 9 | 1284 | 82 | |
More Information
Set Membership: |
|

Krt81
Genomic Map