Assay Details
| Gene Symbol: Ly49i4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009495.1 | NP_001009495.1 | 1 - 2 | 205 | 109 | |
| XM_008763405.1 | XP_008761627.1 | 2 - 3 | 320 | 109 | ||
| GenBank mRNA | AY649834.1 | - | 1 - 2 | 205 | 109 | |
More Information
Set Membership: |
|

Ly49i4
Genomic Map