Assay Details
Gene Symbol: Galr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019173.1 | NP_062046.1 | 2 - 3 | 384 | 100 | |
XM_006241972.2 | XP_006242034.1 | 1 - 2 | 547 | 100 | ||
GenBank mRNA | AF031522.1 | - | 1 - 2 | 359 | 100 | |
AF073798.1 | - | 2 - 3 | 384 | 100 | ||
AF079844.1 | - | 1 - 2 | 359 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
