Assay Details
| Gene Symbol: Clec2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130402.2 | NP_569086.1 | 1 - 2 | 165 | 102 | |
| XM_008763309.1 | XP_008761531.1 | 2 - 3 | 662 | 102 | ||
| GenBank mRNA | AF321552.1 | - | 3 - 4 | 351 | 102 | |
| EF100688.1 | - | 1 - 2 | 165 | 102 | ||
More Information
Set Membership: |
|

Clec2d
Genomic Map