Assay Details
| Gene Symbol: Fadd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152937.2 | NP_690920.1 | 2 | 518 | 86 | |
| GenBank mRNA | AF406779.1 | - | 2 | 518 | 86 | |
| AJ441127.1 | - | 543 | 86 | |||
More Information
Set Membership: |
|

Fadd
Genomic Map