Assay Details
| Gene Symbol: Ugt1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201423.2 | NP_958826.1 | 1 | 524 | 85 | |
| GenBank mRNA | AY435130.1 | - | 1 | 524 | 85 | |
| M34007.1 | - | 1 | 518 | 85 | ||
More Information
Set Membership: |
|

Ugt1a2
Genomic Map