Assay Details
| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108252.1 | NP_001101722.1 | 2 | 1915 | 104 | |
| XM_006256768.2 | XP_006256830.1 | 4 | 2247 | 104 | ||
| XM_008773076.1 | XP_008771298.1 | 3 | 2985 | 104 | ||
More Information
Set Membership: |
|

Usp51
Genomic Map