Assay Details
| Gene Symbol: C1galt1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257487.1 | XP_006257549.1 | 1 - 2 | 155 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257487.1 transcripts.
Set Membership: |
|

C1galt1c1
Genomic Map