Assay Details
| Gene Symbol: Fam8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771560.1 | XP_008769782.1 | 2 - 3 | 853 | 105 | |
| XM_214446.7 | XP_214446.4 | 2 - 3 | 847 | 105 | ||
More Information
Set Membership: |
|

Fam8a1
Genomic Map