Assay Details
| Gene Symbol: Kcnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054000.2 | NP_446452.2 | 2 - 3 | 906 | 73 | |
| GenBank mRNA | M77482.1 | - | 2 - 3 | 909 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Kcnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054000.2 | NP_446452.2 | 2 - 3 | 906 | 73 | |
| GenBank mRNA | M77482.1 | - | 2 - 3 | 909 | 73 | |
Set Membership: |
|