Assay Details
| Gene Symbol: Cd300ld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727841.3 | XP_002727887.2 | 2 - 3 | 468 | 111 | |
| XM_008768422.1 | XP_008766644.1 | 2 - 3 | 701 | 111 | ||
| XM_008768423.1 | XP_008766645.1 | 2 - 3 | 694 | 111 | ||
| XM_008768424.1 | XP_008766646.1 | 2 - 3 | 701 | 111 | ||
More Information
Set Membership: |
|

Cd300ld
Genomic Map