Assay Details
Gene Symbol: Fam196a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143803.1 | NP_001137275.1 | 1 - 2 | 431 | 63 | |
XM_008759969.1 | XP_008758191.1 | 1 - 2 | 564 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143803.1, XM_008759969.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
