Assay Details
| Gene Symbol: Calml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127575.1 | NP_001121047.1 | 4 - 5 | 459 | 62 | |
| XM_006243243.2 | XP_006243305.1 | 3 - 4 | 409 | 62 | ||
| GenBank mRNA | BC158759.1 | - | 4 - 5 | 411 | 7099908 | 62 |
More Information
Set Membership: |
|

Calml4
Genomic Map