Assay Details
| Gene Symbol: NEWGENE_1311521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765485.1 | XP_008763707.1 | 2 - 3 | 662 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1311521| Gene Symbol: NEWGENE_1311521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765485.1 | XP_008763707.1 | 2 - 3 | 662 | 84 | |
Set Membership: |
|