Assay Details
| Gene Symbol: Tbca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013245.1 | NP_001013263.1 | 1 - 2 | 76 | 93 | |
| XM_008760682.1 | XP_008758904.1 | 1 - 2 | 112 | 93 | ||
| GenBank mRNA | BC058155.1 | - | 1 - 2 | 76 | 6888541 | 93 |
More Information
Set Membership: |
|

Tbca
Genomic Map