Assay Details
Gene Symbol: LOC498711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771553.1 | XP_008769775.1 | 4 | 5056 | 67 | |
XM_008771554.1 | XP_008769776.1 | 3 | 5008 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
