Assay Details
| Gene Symbol: NEWGENE_1594649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759095.1 | XP_008757317.1 | 3 | 265 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1594649| Gene Symbol: NEWGENE_1594649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759095.1 | XP_008757317.1 | 3 | 265 | 69 | |
Set Membership: |
|