Assay Details
| Gene Symbol: 31009439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348827.1 | NP_001335769.1 | 1 | 89 | 64 | |
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001179036.1 | NP_011073.1 | 1 | 151 | 64 | |
More Information
Set Membership: |
|

31009439
Genomic Map