Assay Details
| Gene Symbol: SRP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001179882.3 | NP_013018.3 | 1 | 1052 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRP40| Gene Symbol: SRP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001179882.3 | NP_013018.3 | 1 | 1052 | 98 | |
Set Membership: |
|