Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182084.1 | NP_013298.1 | 1 | 1247 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOP56| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182084.1 | NP_013298.1 | 1 | 1247 | 94 | |
Set Membership: |
|