Assay Details
Gene Symbol: MET17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182191.1 | NP_013406.1 | 1 | 1249 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
