Assay Details
| Gene Symbol: CAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182464.1 | NP_013605.1 | 1 | 1352 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAC2| Gene Symbol: CAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182464.1 | NP_013605.1 | 1 | 1352 | 70 | |
Set Membership: |
|