Assay Details
| Gene Symbol: RPS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182934.3 | NP_014303.3 | 1 - 2 | 148 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS7B| Gene Symbol: RPS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001182934.3 | NP_014303.3 | 1 - 2 | 148 | 97 | |
Set Membership: |
|