Assay Details
| Gene Symbol: ETT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001183470.1 | NP_014694.1 | 1 | 1153 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ETT1| Gene Symbol: ETT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001183470.1 | NP_014694.1 | 1 | 1153 | 75 | |
Set Membership: |
|