Assay Details
Gene Symbol: SRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001183847.1 | NP_015292.1 | 1 | 746 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
