Assay Details
| Gene Symbol: NR2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190252.1 | NP_001177181.1 | 2 - 3 | 968 | 66 | |
| NM_001190253.1 | NP_001177182.1 | 2 - 3 | 832 | 66 | ||
| XM_013989100.2 | XP_013844554.1 | 648 | 66 | |||
| GenBank mRNA | AK398528.1 | - | 2 - 3 | 833 | 66 | |
| DN101661.1 | - | 2 - 3 | 643 | 66 | ||
| EU822301.1 | - | 2 - 3 | 833 | 66 | ||
More Information
Set Membership: |
|

NR2F2
Genomic Map