Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013979611.2 | XP_013835065.1 | 3561 | 86 | ||
| XM_013979612.2 | XP_013835066.1 | 3581 | 86 | |||
| XM_021063276.1 | XP_020918935.1 | 2336 | 86 | |||
| XM_021063277.1 | XP_020918936.1 | 3411 | 86 | |||
| XM_021063279.1 | XP_020918938.1 | 1543 | 86 | |||
| GenBank mRNA | AK397751.1 | - | 3 - 4 | 871 | 86 | |
More Information
Set Membership: |
|

TMCC2
Genomic Map