Assay Details
| Gene Symbol: ZFAND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482216.4 | XP_003482264.1 | 3178 | 65 | ||
| XM_003482217.3 | XP_003482265.1 | 1352 | 65 | |||
| XM_005656259.3 | XP_005656316.1 | 737 | 65 | |||
| XM_013988996.2 | XP_013844450.1 | 947 | 65 | |||
| XM_013988999.2 | XP_013844453.1 | 778 | 65 | |||
| XM_013989000.1 | XP_013844454.1 | 640 | 65 | |||
| XM_021098854.1 | XP_020954513.1 | 738 | 65 | |||
| XM_021098855.1 | XP_020954514.1 | 865 | 65 | |||
| XM_021098857.1 | XP_020954516.1 | 781 | 65 | |||
| GenBank mRNA | AK233233.1 | - | 6 - 7 | 757 | 65 | |
| AK235630.1 | - | 5 - 6 | 634 | 65 | ||
| AK393802.1 | - | 5 - 6 | 636 | 65 | ||
More Information
Set Membership: |
|

ZFAND6
Genomic Map