Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244484.2 | NP_001231413.1 | 2 - 3 | 3365 | 76 | |
| GenBank mRNA | AW325610.1 | - | 2 - 3 | 332 | 76 | |
More Information
Set Membership: |
|

PCDH7
Genomic Map