Assay Details
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091639.1 | XP_020947298.1 | 1410 | 75 | ||
| XM_021091640.1 | XP_020947299.1 | 1443 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091639.1 | XP_020947298.1 | 1410 | 75 | ||
| XM_021091640.1 | XP_020947299.1 | 1443 | 75 | |||
Set Membership: |
|