Assay Details
| Gene Symbol: C1H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121415.6 | XP_003121463.3 | 964 | 67 | ||
| XM_021092561.1 | XP_020948220.1 | 1081 | 67 | |||
| GenBank mRNA | AK232997.1 | - | 2 - 3 | 1036 | 67 | |
More Information
Set Membership: |
|

C1H18orf25
Genomic Map