Assay Details
| Gene Symbol: PCF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129708.6 | XP_003129756.2 | 502 | 64 | ||
| XM_013979192.2 | XP_013834646.1 | 472 | 64 | |||
| XM_021062577.1 | XP_020918236.1 | 500 | 64 | |||
| XM_021062578.1 | XP_020918237.1 | 473 | 64 | |||
| GenBank mRNA | AK236905.1 | - | 1 - 2 | 473 | 64 | |
More Information
Set Membership: |
|

PCF11
Genomic Map