Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929397.5 | XP_001929432.1 | 927 | 58 | ||
| XM_003355137.3 | XP_003355185.1 | 2044 | 58 | |||
| XM_005663337.3 | XP_005663394.1 | 926 | 58 | |||
| XM_005663338.3 | XP_005663395.1 | 482 | 58 | |||
| XM_013997033.1 | XP_013852487.1 | 333 | 58 | |||
| XM_021089743.1 | XP_020945402.1 | 405 | 58 | |||
| XM_021089744.1 | XP_020945403.1 | 482 | 58 | |||
| GenBank mRNA | AK393748.1 | - | 3 - 4 | 370 | 58 | |
| AK394898.1 | - | 2 - 3 | 791 | 58 | ||
More Information
Set Membership: |
|

SHC1
Genomic Map