Assay Details
| Gene Symbol: MMUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214405.1 | NP_999570.1 | 3 - 4 | 867 | 111 | |
| GenBank mRNA | AJ496801.1 | - | 3 - 4 | 867 | 111 | |
| AK343989.1 | - | 3 - 4 | 892 | 111 | ||
| AK389866.1 | - | 4 - 5 | 865 | 111 | ||
| AK396479.1 | - | 3 - 4 | 954 | 111 | ||
More Information
Set Membership: |
|

MMUT
Genomic Map