Assay Details
| Gene Symbol: GTF2H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123120.1 | NP_001116592.1 | 7 - 8 | 743 | 78 | |
| XM_005665731.3 | XP_005665788.1 | 997 | 78 | |||
| XM_005665732.3 | XP_005665789.1 | 857 | 78 | |||
More Information
Set Membership: |
|

GTF2H4
Genomic Map