Assay Details
| Gene Symbol: RPS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214363.1 | NP_999528.1 | 3 - 4 | 203 | 59 | |
| GenBank mRNA | AK234793.1 | - | 1398 | 59 | ||
| AK236805.1 | - | 3 - 4 | 215 | 59 | ||
| AK344762.1 | - | 3 - 4 | 213 | 59 | ||
| AK392759.1 | - | 3 - 4 | 214 | 59 | ||
| AY610355.1 | - | 3 - 4 | 201 | 59 | ||
| X79417.1 | - | 203 | 59 | |||
More Information
Set Membership: |
|

RPS12
Genomic Map