Assay Details
| Gene Symbol: EIF4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100195.1 | NP_001093665.1 | 4 - 5 | 381 | 82 | |
| XM_021068176.1 | XP_020923835.1 | 621 | 82 | |||
| GenBank mRNA | AK231721.1 | - | 4 - 5 | 389 | 82 | |
| AK391718.1 | - | 4 - 5 | 381 | 82 | ||
| DQ351285.1 | - | 4 - 5 | 381 | 82 | ||
More Information
Set Membership: |
|

EIF4A2
Genomic Map