Assay Details
| Gene Symbol: CPEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134043.5 | XP_003134091.2 | 3340 | 67 | ||
| XM_003359814.4 | XP_003359862.1 | 3284 | 67 | |||
| XM_005657622.3 | XP_005657679.1 | 3328 | 67 | |||
| XM_005657623.3 | XP_005657680.1 | 3316 | 67 | |||
| XM_005657625.3 | XP_005657682.1 | 3270 | 67 | |||
| GenBank mRNA | AK394633.1 | - | 6 - 7 | 741 | 67 | |
More Information
Set Membership: |
|

CPEB4
Genomic Map